Learn More
Medchemexpress LLC CCT369260 | 2647503-57-7 | 99.7% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY129188100MG
Encompass_Preferred
CCT369260 is a chemical probe and an orally active B-cell lymphoma 6 (BCL6) inhibitor. It demonstrates anti-tumor activity and exhibits an IC50 of 520 nM.
  • Orally active B-cell lymphoma 6 (BCL6) inhibitor
  • Exhibits anti-tumor activity
  • IC50 of 520 nM against BCL6
  • Significantly inhibits BCL6 in OCI-Ly1 DLBCL xenograft model (in vivo study)
Catalog No. 50-003-39363