Learn More
Medchemexpress LLC CCT374705 | 2640647-90-9 | C21H18ClF3N4O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY153521100MG
Encompass_Preferred
CCT374705 is an orally active BCL6 inhibitor (IC50 = 4.8 nM) with potent antiproliferative effects in vitro. It effectively inhibits tumor growth in a lymphoma xenograft mouse model.
  • Effectively inhibits tumor growth in a lymphoma xenograft mouse model.
  • Good oral bioavailability.
  • Low total clearance and plasma protein binding, leading to high free concentrations in Balb/C mice.
  • Modulates BCL6 activity.
Catalog No. 50-003-72766