Learn More
AdipoGen CCVJ
SDP
Supplier:  AdipoGen CDXC0085M010
Encompass_Preferred
Chemical. CAS 142978-18-5. Formula C16H16N2O2. MW 268.31. Synthetic. Fluorescent probe for binding to proteins etc. quantum yield increases by decreasing free rotation. Fluorescent molecular rotor.
Catalog No. 50-256-1484