Learn More
TARGETMOL CHEMICALS INC CCX354 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T152535MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CCX354 (CCR1 antagonist 1) is a potent small molecule CCR1 antagonist with anti-inflammatory activity for the treatment of rheumatoid arthritis. purity: 99%
Catalog No. 50-258-1240