Learn More
Apexbio Technology LLC CD 1530 107430-66-0 10mg
SDP
Supplier:  Apexbio Technology LLC B710210
Encompass_Preferred
CD1530 (CAS 107430-66-0) is a selective retinoic acid receptor (RAR) agonist targeting primarily RAR with lower affinity for RAR and RAR It is designed to activate RAR -mediated pathways thereby regulating cellular growth and apoptosis CD1530 exerts its biological activity by selectively activating RAR In transcriptional assays CD1530 demonstrates potent activation of RAR with an AC50 of approximately 1 8 nM It also inhibits the CYP26A1 enzyme (IC50 530 nM) Based on these pharmacological properties CD1530 holds research potential in preclinical cancer studies particularly in oral cavity tumorigenesis models and investigations of retinoid signaling and metabolism
Catalog No. 50-000-90902