Learn More
ABclonal Technology CD10/MME Rabbit mAb
SDP
Supplier:  ABclonal Technology A23509100UL
Encompass_Preferred
The protein encoded by this gene is a type II transmembrane glycoprotein and a common acute lymphocytic leukemia antigen that is an important cell surface marker in the diagnosis of human acute lymphocytic leukemia (ALL) The encoded protein is present on leukemic cells of pre-B phenotype which represent 85% of cases of ALL This protein is not restricted to leukemic cells however and is found on a variety of normal tissues The protein is a neutral endopeptidase that cleaves peptides at the amino side of hydrophobic residues and inactivates several peptide hormones including glucagon enkephalins substance P neurotensin oxytocin and bradykinin
Catalog No. 50-000-06433