Learn More
Medchemexpress LLC CD117/c-Kit Mouse 50ug
SDP
Supplier:  Medchemexpress LLC HYP7288050UG
Encompass_Preferred
CD117/c-Kit protein is a tyrosine protein kinase that acts as a cell surface receptor for KITLG/SCF and regulates cell survival proliferation hematopoiesis stem cell maintenance gametogenesis mast cell development migration function and melanogenesis CD117/c-Kit Protein Mouse (HEK293 His) is the recombinant mouse-derived CD117/c-Kit protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-81833