Learn More
Medchemexpress LLC CD1530 | 107430-66-0 | 98.01% | 398.49 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY108527100MG
Encompass_Preferred
CD1530 is an orally active, selective RARγ agonist and an antibacterial agent. It works by reducing Smad1/5/8 phosphorylation and overall Smad levels, as well as decreasing β-catenin, MMP9 protein, and Reactive Oxygen Species (ROS) levels. It has shown efficacy in inhibiting heterotopic ossification, promoting Achilles tendon healing, and inhibiting muscle fatty infiltration. It is suitable for research in orthopedic diseases, such as heterotopic ossification and Achilles tendon injury, and muscle diseases, including those related to muscle fatty infiltration.
  • Orally active and selective RARγ agonist
  • Functions as an antibacterial agent
  • Reduces Smad1/5/8 phosphorylation and overall Smad levels
  • Decreases β-catenin, MMP9 protein, and ROS levels
  • Inhibits heterotopic ossification
  • Promotes Achilles tendon healing
  • Inhibits muscle fatty infiltration
Catalog No. 50-003-46205