Learn More
Beckman Coulter CD159a-PE, Z199, 1 mL
Supplier:  Beckman Coulter IM3291U
Encompass
CD159a (NKG2a) is a 40 kDa (reduced), 75 kDa (unreduced) type II transmembrane protein associated with CD94 to form a disulphide-linked heterodimer. CD159a is a member of the Ca++-dependent (C-type) lectin family.
Catalog No. NC9086290