Learn More
Medchemexpress LLC CD19 Mouse Biotiny 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7752750UG
Encompass_Preferred
The CD19 protein acts as a coreceptor for B cell antigen receptors lowering the signaling threshold and initiating B cell responses to antigens It activates a cascade leading to PI3K activation and Ca(2 ) mobilization CD19 Protein Mouse (Biotinylated HEK293 His) is the recombinant mouse-derived CD19 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-87111