Learn More
Sino Biological Recombinant Human CD19 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11880H08HB100
Encompass_Preferred
A DNA sequence encoding the Human CD19 (P15391) (Met1-Lys291) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6236