Learn More
Medchemexpress LLC CD1C-B2M heterodimer protein, rhesus macaque (HEK293, His) | >95.0% | 10 UG
SDP
Supplier:  Medchemexpress LLC HYP7621010UG
Encompass_Preferred
Recombinant CD1C-B2M heterodimer derived from rhesus macaque, expressed in HEK293 cells and supplied lyophilized from PBS (pH 7.4). The protein contains a C-terminal 6xHis tag, has >95.0% purity by reducing SDS-PAGE, and an approximate molecular weight of 44.7 kDa (33.1 + 11.6 kDa subunits). Store frozen and aliquot for long-term stability.
  • Recombinant dimeric protein suitable for assay development and characterization.
  • Expressed in HEK293 for eukaryotic post-translational modifications.
  • C-terminal 6xHis tag enables affinity purification and detection.
  • Lyophilized formulation for extended shelf life and convenient storage.
  • High purity (>95.0%) ensures consistency in downstream experiments.
  • Stable when aliquoted and stored at -20°C or -80°C for long-term use.
Catalog No. 50-002-91846