Learn More
Cayman Chemical CD1d EXrAClular DomaIn2 100ug
SDP
Supplier:  Cayman Chemical 34877100 UG
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged CD1d extracellular domain and β2-microglobulin expressed in HEK293 cells • Amino acids: 20-301 (CD1d), 21-119 (β2-microglobulin) • MW: 33 (CD1d) and 12 (β2-microglobulin) kDa
Catalog No. 50-291-0521