Learn More
KACTUSBIO INC Human CD20 Protein-VLP (CD2-HM122) . 1EA=100UG.
Supplier:  KACTUSBIO INC CD2HM122
Encompass
Recombinant Human CD20 Protein-VLP is expressed from HEK293.It contains Met1-Pro297.
Catalog No. NC2956817