Learn More
Genetex Inc CD20 Monoclonal Antibody [MSVA-020R] HistoMAX™ (GTX04392) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04392
Encompass_Preferred
The CD20 antibody [MSVA-020R] HistoMAX™ (GTX04392) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human mucosa from appendix tissue using GTX04392 CD20 antibody [MSVA-020R] HistoMAX. Strong CD20 staining of a germinal centre in the appendix mucosa while epithelial cells are completely negative.
Catalog No. NC3666745