Learn More
CAPRICO BIOTECHNOLOGIES INC CD20 PE Antibody, 4037025
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4037025
Encompass_Preferred
The Caprico CD20 PE Antibody is a mouse monoclonal antibody that binds to human CD20. It is conjugated to R-phycoerythrin (PE) and is intended for flow cytometry applications. This antibody recognizes a 35-37 kDa protein expressed on the surface of B lymphocytes, from early pre-B to mature B cells. It does not react with plasma cells or stem cells. This antibody can be used for immunophenotyping B cells in normal and malignant tissues. It is available in 100 tests of 50 μL volume.
Catalog No. 50-227-9064