Learn More
Enzo Life Sciences DEC-205 (mouse) monoclonal antibody (NLDC-145) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS1220100
Encompass_Preferred
Clone: NLDC-145. Isotype: IgG2a. Immunogen: Mouse lymphoid tissue stroma. Species reactivity: Mouse. Specificity: Recognizes mouse dendritic cells. Applications: Flow Cytometry, IF, IHC. Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In TRIS-NaCl, pH 8.
Catalog No. 50-201-2684