Learn More
Medchemexpress LLC CD206/MMR Human HE 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7873650UG
Encompass_Preferred
The CD206/MMR protein acts as a multifunctional mediator in macrophages promoting glycoprotein internalization It has a broad binding capacity interacting with both sulfated and non-sulfated polysaccharide chains CD206/MMR Protein Human (HEK293 His) is the recombinant human-derived CD206/MMR protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-95052