Learn More
Cayman Chemical CD226 EXrAClular DomaInh 100ug
SDP
Supplier:  Cayman Chemical 31837100 UG
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged CD226 expressed in HEK293 cells • Amino acids: 19-247 • MW: 27.6 kDa
Catalog No. 50-294-8643