Learn More
Cayman Chemical CD226 EXrAClular DomaInm 100ug
SDP
Supplier:  Cayman Chemical 32072100 UG
Encompass_Preferred
Source: Active recombinant mouse C-terminal His-tagged CD226 expressed in HEK293 cells • Amino acids: 19-254 • MW: 28.2 kDa
Catalog No. 50-294-9395