Learn More
Medchemexpress LLC CD23/Fc epsilon RII 5ug
SDP
Supplier:  Medchemexpress LLC HYP743205UG
Encompass_Preferred
CD23 also known as Fc epsilon Receptor II (Fc epsilon RII) is a low-affinity receptor for immunoglobulin E (IgE) and complement receptor 2 (CR2/CD21) CD23 regulates IgE production and B-cell differentiation aiding in IGE-dependent antigen uptake CD23/Fc epsilon RII Protein Rat (HEK293 Fc) is the recombinant rat-derived CD23/Fc epsilon RII protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-78805