Learn More
Genscript Corporation CD27/TNFRSF7 hFc ChimeraBioti
Supplier:  Genscript Corporation Z04062500
Encompass
CD27 also known as TNFRSF7 is an approximately 55 kDa transmembrane protein in the TNF receptor superfamily It functions as a co stimulatory molecule that supports lymphocyte activation and survival It binds to ligand CD70 and plays a key role in regulating B-cell activation and immunoglobulin synthesis
Catalog No. 50-000-58160