Learn More
ABclonal Technology CD298/ATP1B3 Rabbit PolymAb
SDP
Supplier:  ABclonal Technology A19637PM20UL
Encompass_Preferred
The protein encoded by this gene belongs to the family of Na K and H K ATPases beta chain proteins and to the subfamily of Na K -ATPases Na K -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane These gradients are essential for osmoregulation for sodium-coupled transport of a variety of organic and inorganic molecules and for electrical excitability of nerve and muscle This enzyme is composed of two subunits a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta) The beta subunit regulates through assembly of alpha beta heterodimers the number of sodium pumps transported to the plasma membrane The glycoprotein subunit of Na K -ATPase is encoded by multiple genes This gene encodes a beta 3 subunit This gene encodes a beta 3 subunit A pseudogene exists for this gene and it is located on chromosome 2
Catalog No. 50-000-08189