Learn More
Diagnostic Biosystems CD3 CD3 LN10
Supplier:  Diagnostic Biosystems PDM186
Encompass_Preferred
The Clone LN10 is specific for the non-glycosylated epsilon chain of the human CD3 molecule. Clone LN10 recognizes T cells in thymus, bone marrow, peripheral lymphoid tissue and blood and is a pan T cell marker. The CD3 molecule consists of five different polypeptide chains with molecular weights ranging from 16 to 28 kD. The CD3 antigen is first detected in early thymocytes and its appearance probably represents one of the earliestsigns of commitment to the T cell lineage.
Catalog No. 50-240-7567