Learn More
Newcomer Supply CD3, T-Cell Control Slides, 10/set
SDP
Supplier:  Newcomer Supply 3770A
Encompass
Newcomer Supply CD3, T-Cell Control Slides are for the positive immunohistochemical staining of T-Cells. CD3 is considered to be a pan-T-cell marker, and widely used in detection of T-cell malignancies, both immature and mature. CD3, T-Cell Control Slides contain sections of positive staining tonsil.
Catalog No. NC2101952