Learn More
TARGETMOL CHEMICALS INC CD38 INHIBITOR 1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T149135MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CD38 inhibitor 1 is an inhibitor of CD38. For hCD38 and mouse CD38 the IC50s are 7.3 nM and 1.9 nM. purity: 99%
Catalog No. 50-257-7371