Learn More
Apexbio Technology LLC CD38 inhibitor 1(Synonyms: 78c, CD38-IN-78c, CD38 inhibitor 78c, CD38 selective inhibitor), 5mg, CAS: 1700637-55-3.
SDP
Supplier:  Apexbio Technology LLC B87525
Encompass_Preferred
CD38 inhibitor 1 (CAS 1700637-55-3) is a potent selective small molecule inhibitor targeting CD38 enzymatic activity in human and mouse systems Reported IC50 values are approximately 7 3 nM for human CD38 and 1 9 nM for mouse CD38 CD38 is implicated in intracellular calcium signaling and NAD metabolic regulation Blocking CD38 enzymatic function enables exploration of its biological roles and potential therapeutic relevance in immunological processes inflammation autoimmune disorders and cancer metabolism through both in vitro and in vivo studies
Catalog No. 50-000-60401