Learn More
Medchemexpress LLC CD3D-CD3E Heterodime 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7680250UG
Encompass_Preferred
The CD3D protein is an important component of the TCR-CD3 complex on T lymphocytes and is critical for adaptive immune responses After APC activates TCR CD3D together with CD3E CD3G and CD3Z transmits signals through ITAM and activates downstream pathways CD3D-CD3E Heterodimer Protein Rhesus Macaque (HEK293 hFc) is a recombinant protein dimer complex containing Rhesus Macaque-derived CD3D-CD3E Heterodimer protein expressed by HEK293 with N-hFc
Catalog No. 50-001-85788