Learn More
Biotium CD3e, Mouse(145-2C11), CF640R conjugate, 0.1mg/mL
Supplier:  Biotium BNC402013100
Encompass_Preferred
CD3 chains. CD3 plays a critical role in TCR signal transduction, T-cell activation, and antigen recognition by binding the peptide/MHC antigen complex. The 145-2C11 antibody is useful for in vitro blocking and activation assays, as well as apoptosis induction and in vivo T-cell depletion.Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. CF Dyes offer exceptional brightness and photostability. Note: Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.
Catalog No. 50-006-47800