Learn More
Genetex Inc CD4 Monoclonal Antibody [MSVA-004R] HistoMAX™ (GTX04379) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04379
Encompass_Preferred
The CD4 antibody [MSVA-004R] HistoMAX™ (GTX04379) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human B-cell chronic lymphocytic leukemia (B-CLL) tissue using GTX04379 CD4 antibody [MSVA-004R] HistoMAX. B CLL containing numerous CD4 positive lymphocytes.
Catalog No. NC3666286