Learn More
Biotium CD4 (T-Helper/Inducer Cell Marker)(CD4/3027), CF405S conjugate, 0.1mg/mL
Supplier:  Biotium BNC043027100
Encompass_Preferred
The immunogen for this Mab is derived from the extracellular domain of CD4, a membrane glycoprotein of T lymphocytes that interacts with major histocompatibility complex class II antigens and is also a receptor for the human immunodeficiency virus. This protein is expressed not only in T lymphocytes, but also in B cells, macrophages, and granulocytes. It is also expressed in specific regions of the brain. The protein functions to initiate or augment the early phase of T-cell activation, and may function as an important mediator of indirect neuronal damage in infectious and immune-mediated diseases of the central nervous system. The majority of peripheral T-cell lymphomas are derived from the T-helper/regulatory cell subset so that most mature T-cell neoplasms are CD4 /CD8-. Anti-CD4 is used in the immunohistochemical staining of lymphoproliferative disorders to evaluate tumors with CD4 aberrant expression. Primary antibodies are available purified, or with a selection of fluo
Catalog No. 50-006-17301