Learn More
Enzo Life Sciences CD40 (human) monoclonal antibody (mAb 89) (Alexa Fluor 647) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS1400100
Encompass_Preferred
Clone: mAb 89. Isotype: IgG1. Immunogen: Anti-μ activated tonsillar human B cells. Species reactivity: Human. Applications: Flow Cytometry, IF. Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-201-2701