Learn More
Genetex Inc CD45 Monoclonal Antibody [MSVA-045R] HistoMAX™ (GTX04432) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04432
Encompass_Preferred
The CD45 antibody [MSVA-045R] HistoMAX™ (GTX04432) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human diffuse large B-cell lymphoma (DLBCL) from lymph node tissue using GTX04432 CD45 antibody [MSVA-045R] HistoMAX. Diffuse large B cell lymphoma showing strong CD45 staining of tumor cells.
Catalog No. NC3666940