Learn More
Genetex Inc CD45RA Monoclonal Antibody [MSVA-472M] HistoMAX™ (GTX04372) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04372
Encompass_Preferred
The CD45RA antibody [MSVA-472M] HistoMAX™ (GTX04372) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human thymus tissue using GTX04372 CD45RA antibody [MSVA-472M] HistoMAX. More than 99 of the cortical thymocytes are CD45RA negative. The rate of CD45RA positive cells is markedly higher in the medulla.
Catalog No. NC3664120