Learn More
Medchemexpress LLC CD47 Human HEK293 10ug
SDP
Supplier:  Medchemexpress LLC HYP733510UG
Encompass_Preferred
CD47 Protein Human (HEK293 Fc) is a polypeptide chain with the C-terminal human IgG1 Fc fragment produced in HEK293 cells CD47 is a prominent target in cancer immunotherapy
Catalog No. 50-001-79827