Learn More
Medchemexpress LLC CD47 Human HEK293 10ug
SDP
Supplier:  Medchemexpress LLC HYP7291710UG
Encompass_Preferred
CD47 is an adhesion protein that regulates integrin signaling through G protein activation and serves as a receptor for thrombospondin THBS1 affecting signal transduction cardiovascular homeostasis inflammation apoptosis angiogenesis self-renewal and Immunomodulatory It regulates pulmonary endothelin EDN1 signaling acts as a pressor in blood pressure regulation and is critical for memory formation CD47 Protein Human (HEK293) is the recombinant human-derived CD47 protein expressed by HEK293 with tag free
Catalog No. 50-001-82896