Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CD47 protein, rat (HEK293, Fc) | 95.0% | 500 UG
SDP

Supplier:  Medchemexpress LLC HYP75402500UG

Encompass_Preferred

Recombinant rat CD47 extracellular domain expressed in HEK293 cells and supplied as a C-terminal human Fc-tagged fusion protein. Provided as a purified reagent for biochemical and cell-based research, it is suitable for binding studies, ELISA capture/detection, and receptor-ligand interaction assays. Supplied frozen; estimated purity ~95%.

  • Recombinant rat CD47 extracellular domain with C-terminal human Fc tag.
  • Expressed in HEK293 cells for mammalian-type glycosylation.
  • Estimated purity 95.0%.
  • Stored frozen at -20°C.
  • Suitable for binding assays, ELISA capture/detection, and receptor interaction studies.
  • Available in microgram-scale quantities (e.g., 500 UG).

Catalog No. 50-002-93425


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.