Learn More
Medchemexpress LLC CD5 Human Biotinyl 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7809350UG
Encompass_Preferred
The CD5 protein is a potential receptor that regulates T cell proliferation Its interactions with CD72/LYB-2 and PTPN6/SHP-1 indicate multifaceted roles in cellular processes serving as key mediators of T cell responses CD5 Protein Human (Biotinylated HEK293 Fc-Avi) is the recombinant human-derived CD5 protein expressed by HEK293 with C-Avi C-hFc labeled tag
Catalog No. 50-001-87527