Learn More
ABclonal Technology CD58 Rabbit mAb
SDP
Supplier:  ABclonal Technology A22276100UL
Encompass_Preferred
This gene encodes a member of the immunoglobulin superfamily The encoded protein is a ligand of the T lymphocyte CD2 protein and functions in adhesion and activation of T lymphocytes The protein is localized to the plasma membrane Alternatively spliced transcript variants have been described
Catalog No. 50-000-05006