Learn More
Medchemexpress LLC CD5L Mouse HEK293 5ug
SDP
Supplier:  Medchemexpress LLC HYP727175UG
Encompass_Preferred
The CD5L protein is primarily expressed by macrophages and regulates inflammatory responses in infection or atherosclerosis CD5L is incorporated into adipocytes via CD36-mediated endocytosis thereby inhibiting lipid droplet size and affecting lipolysis CD5L Protein Mouse (HEK293 His) is the recombinant mouse-derived CD5L protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-82321