Learn More
Medchemexpress LLC CD64 Rabbit HEK293 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7732450UG
Encompass_Preferred
The CD64 protein is a high-affinity receptor for the immunoglobulin gamma Fc region and plays a key role in innate and adaptive immune responses It prevents FCGR1A degradation by interacting with FCERG1 and FLNA to form a functional signaling complex CD64 Protein Rabbit (HEK293 His) is the recombinant Rabbit-derived CD64 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86907