Learn More
ABclonal Technology CD66a/CEACAM1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2547520UL
Encompass_Preferred
Enables several functions including signaling receptor binding activity virion binding activity and virus receptor activity Involved in several processes including common myeloid progenitor cell proliferation granulocyte colony-stimulating factor signaling pathway and negative regulation of macromolecule metabolic process Acts upstream of or within several processes including modulation by host of viral process regulation of T cell activation and regulation of intracellular signal transduction Located in cell projection membrane external side of plasma membrane and extracellular space Is expressed in several structures including alimentary system central nervous system extraembryonic component genitourinary system and skeleton Orthologous to several human genes including CEACAM1 (CEA cell adhesion molecule 1)
Catalog No. 50-000-13028