Learn More
ABclonal Technology CD66a/CEACAM1 Rabbit PolymAb
SDP
Supplier:  ABclonal Technology A26413PM100UL
Encompass_Preferred
This gene encodes a protein from the carcinoembryonic antigen (CEA) family part of the immunoglobulin superfamily It is located within a 1 2 Mb cluster on chromosome 19 with CEA cell adhesion molecules and pregnancy-specific glycoproteins The protein initially described as biliary glycoprotein in bile ducts is a cell-cell adhesion molecule found on leukocytes epithelia and endothelia It mediates cell adhesion through homophilic and heterophilic binding This protein is involved in tissue differentiation three-dimensional structure angiogenesis apoptosis tumor suppression metastasis and immune response modulation Multiple transcript variants encoding different isoforms have been identified though the full-length nature of all variants is unclear
Catalog No. 50-000-10423