Learn More
ABclonal Technology CD69 Rabbit mAb
SDP
Supplier:  ABclonal Technology A22171100UL
Encompass_Preferred
This gene encodes a member of the calcium dependent lectin superfamily of type II transmembrane receptors Expression of the encoded protein is induced upon activation of T lymphocytes and may play a role in proliferation Furthermore the protein may act to transmit signals in natural killer cells and platelets
Catalog No. 50-000-05432