Learn More
ACROBiosystems Recombinant Protein;Human CD72 Protein, Fc Tag;HEK293;1MG;CD2,CD72,Lyb-2,Lyb2
Supplier:  ACROBiosystems CD2H52511MG
Encompass_Preferred
Recombinant Protein;Human CD72 Protein, Fc Tag;1MG;Human CD72, Fc Tag (CD2-H5251) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Accession # AAH30227).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 54 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8487