Learn More
TRIBIOSCIENCE INC 5’-Nucleotidase CD73 Activity Colorimetric Assay, 100 Tests
SDP
Supplier:  TRIBIOSCIENCE INC TBS2046
Encompass_Preferred
Tribioscience’s CD73 Activity Kit provides a simple, easy assay that relies on the quantification of free phosphate with the Malachite Green complex. In this assay, the action of CD73 on the substrate generates free phosphate, which combines malachite green to form color complex. The color intensity is quantified at 620 nm. The assay kit also includes CD73 enzyme as positive control.
Catalog No. NC3454574