Learn More
Medchemexpress LLC CD73-IN-3 | 2375815-63-5 | 99.89% | 286.33 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13724650MG
Encompass_Preferred
CD73-IN-3 is a potent CD73 inhibitor with an IC50 of 7.3 nM in a Calu6 human cell assay. This compound, derived from patent WO2019168744 A1, demonstrates potential utility in cancer research. In vitro studies show it effectively inhibits CD73 activity in human serum with an EC50 of 0.213 μM.
  • Potent CD73 inhibitor with an IC50 of 7.3 nM in Calu6 human cell assays.
  • Inhibits CD73 activity in human serum with an EC50 of 0.213 μM.
  • Shows inhibition rates of 56.8%, 71.7%, and 76.9% at concentrations of 0.52 μM, 1.56 μM, and 4.68 μM.
  • Potential for cancer research.
Catalog No. 50-003-49127