Learn More
Medchemexpress LLC CD74 Human HEK293 50ug
SDP
Supplier:  Medchemexpress LLC HYP7293150UG
Encompass_Preferred
CD74 is a key component of the class II major histocompatibility complex and serves as a key chaperone in antigen presentation in the regulation of immune responses It doubles as a cell surface receptor for macrophage migration inhibitory factor (MIF) initiating survival pathways and cell proliferation CD74 Protein Human (HEK293 His) is the recombinant human-derived CD74 protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-82724