Learn More
Medchemexpress LLC CD79B Human Biotin 20ug
SDP
Supplier:  Medchemexpress LLC HYP7076820UG
Encompass_Preferred
Recombinant human CD79B extracellular domain, biotinylated via an Avi tag and produced in HEK293 cells with a C-terminal His tag. This product is intended for use in binding assays, ELISA, receptor interaction studies, and other immunology research requiring a biotinylated B-cell receptor component reagent.
  • Biotinylated via Avi tag for streptavidin-based capture and detection.
  • Expressed in mammalian cells to preserve native post-translational modifications.
  • Includes C-terminal His tag to enable purification and detection.
  • Suitable for binding assays, ELISA, and receptor interaction studies.
  • Provided as a purified recombinant protein for research use.
Catalog No. 50-001-97123