Learn More
Medchemexpress LLC CD79B Human HEK293 5ug
SDP
Supplier:  Medchemexpress LLC HYP705415UG
Encompass_Preferred
Recombinant human CD79B protein encompassing residues A29-D159, expressed in HEK293 cells and engineered with a C-terminal 6xHis tag. Supplied in microgram quantities for use as an antigen, assay reagent, or control in biochemical and cell-based studies of B-cell receptor signaling, internalization, and antigen presentation. The mammalian expression system helps preserve native post-translational modifications relevant to function.
  • Recombinant human sequence covering A29-D159 (UniProt P40259).
  • Expressed in HEK293 to retain mammalian post-translational modifications.
  • C-terminal 6xHis tag for straightforward purification and detection.
  • Available in microgram sizes suitable for biochemical and immunoassays.
  • Intended for studies of B-cell receptor signaling and antigen presentation.
Catalog No. 50-001-85514